C6 NBD-Ceramide (d18:1/6:0)
Dodavatel: Cayman Chemical
Synonyms:
NBD C6-Ceramide, N-[7-(4-nitrobenzo-2-oxa-1,3-diazole)]-6-aminocaproyl-D-erythro-sphingosine, 6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosine, N-C6-NBD-D-erythro-Sphingosine
62527-1
62527-10
62527-5
CAYM62527-1EA
7240
CZK
CAYM62527-1
CAYM62527-10
CAYM62527-5
C6 NBD-Ceramide (d18:1/6:0)
C6 NBD-Ceramide
Vzorec:
C₃₀H₄₉N₅O₆ MW: 575,74 g/mol Storage Temperature: Freezer |
CAS číslo:
94885-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...