Tisk…
Respirátory částicové FFP1/FFP2/FFP3, řady Aura™ 9300+Gen3
Dodavatel: 3M
9322PLUSGEN3SV 9332PLUSGEN3SV 9322PLUSGEN3 9332PLUSGEN3
111-2393EA 834 CZK
111-2393 111-2389 111-2395 111-2391
Respirátory částicové FFP1/FFP2/FFP3, řady Aura™ 9300+Gen3
Respirators Polomasky filtrační, jednorázové Polomasky k filtraci částic
Aura™ particulate respirator 9300+Gen3 series is the third generation of the best-selling 3M™ Aura™ flat-fold respirator.
  • Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
  • New 3M™ Cool Flow™ Comfort valve allows a greater percentage of warm, exhaled air to escape, helping to increase comfort (compared to 3M™ Aura™ 9300+ equivalent model)
  • 3M Advanced electret (filter) media helps you breathe easier
  • Innovative tabs on the upper and lower panels simplify positioning and adjustment
  • Easy to store away with flat-fold design and individual wrapping
  • Available in standard packs (20 per carton) or small packs (5 per carton)
  • Made in the UK
Get comfortable and reliable FFP2 and FFP3 breathing protection in hazardous work environments, with 3M™ Aura™ disposable respirator 9300+Gen3 series. This is the third generation of the best selling 3M Aura flat-fold particulate respirator, featuring the original 3-panel, flat-fold design. This range of particulate respirators are made using 3M Advanced Electret (filter) Media (AEM), a high performance material that’s been engineered for easy, unrestricted breathing. The range includes the 3M™ Cool Flow™ comfort valve, which opens more easily and for longer than the original 3M™ Cool Flow™ valve.
Innovative tabs on the upper and lower panels make wearing, taking off, or adjusting that bit easier (especially in combination with the new grip feature on valved models). The braided, colour-coded headbands help improve durability and allow quick identification the protection level offered: blue for FFP2 (NPF 12) and red for FFP3 (NPF 50). The soft nose foam is comfortable on skin, and an embossed, sculpted upper panel helps reduce eyewear fogging and works well with 3M eyewear products. An integrated nose clip helps achieve a comfortable fit to a broad range of face shapes.
One size fits all.