Tisk…
Poly-L-lysine hydrobromide
Dodavatel: Merck
P1524-100MG P1524-1G P1524-25MG P1524-500MG
SIALP1524-100MGEA 6820 CZK
SIALP1524-100MG SIALP1524-1G SIALP1524-25MG SIALP1524-500MG
Poly-L-lysine hydrobromide
Proteiny
Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water.
Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations.