10-Acetyl-3,7-dihydroxyphenoxazine ≥98%
Dodavatel: Cayman Chemical
|
Varování
|
Synonyms:
1-(3,7-Dihydroxy-10H-phenoxazin-10-yl)ethanone, ADHP, Amplex® Red (Registered trademark of LifeTechnologies)
10-Acetyl-3,7-dihydroxyphenoxazine (ADHP) is a highly sensitive, stable substrate for horseradish peroxidase (HRP) that enables selective detection of H₂O₂.
This non-fluorescent reagent reacts with H₂O₂ to produce the fluorescent compound resorufin, which can be analysed using an excitation wavelength of 520 to 550 nm and an emission wavelength of 585 to 595 nm. In a 96 well plate format, ADHP enables detection of H₂O₂ at a concentration as low as 5 pmol per 100 µl sample.
Vzorec:
C₁₄H₁₁NO₄ MW: 257,24 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00467861 CAS číslo: 119171-73-2 |
Výsledky specifikačního testu
Purity | ≥98% |
λmax | 250, 285 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...