Tisk…
Flame-retardant trousers, Flamestat 2148 ATHS
Dodavatel: FRISTADS KANSAS
12503854058 125038540D116 12503854060 12503854048 12503854054 12503854044 125038540D84 125038540D96 125038540D104 125038540D112 12503854052 12503854062 12503854050 12503854046 12503854056 125038540D88 125038540D92 125038540D100 125038540D108
KWIN12503854058EA 5580 CZK
KWIN12503854058 KWIN125038540D116 KWIN12503854060 KWIN12503854048 KWIN12503854054 KWIN12503854044 KWIN125038540D84 KWIN125038540D96 KWIN125038540D104 KWIN125038540D112 KWIN12503854052 KWIN12503854062 KWIN12503854050 KWIN12503854046 KWIN12503854056 KWIN125038540D88 KWIN125038540D92 KWIN125038540D100 KWIN125038540D108
Flame-retardant trousers, Flamestat 2148 ATHS
Pracovní oděvy
Dirt, oil and water repellent trousers with inherent flame protection adapted for ATEX environments with no metal details. Manufactured from 45% modacrylic, 35% cotton, 18% polyamide and 2% anti-static fibre. Approved for industrial laundry.
  • Two front pockets; D-ring at waistband
  • Two back pockets with flap and velcro fastening
  • Leg pocket with flap and snap fastening
  • Leg pocket with flap and concealed snap fastening, phone pocket with flap and velcro fastening, D-ring
  • Adjustable leg length with 5 cm hem allowance
Weight: 300 g/m²
Certifikace: EN 6148More Product Information
Objednat
Produktová specifikace
Model 2148 ATHS
Environmentally Preferable
Aplikace oblečení Welding,Industrial
Risk area Flame-retardant,Anti-static
Rod Pro muže
Sleeve or leg length Long
Tkanina 45% modacrylic, 35% cotton, 18% polyamide and 2% anti-static fibre
Hmotnost 300 g/m²