Tisk…

PTFE, free flowing powder 1 µm, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Polytetrafluoroethylene, Teflon

M-Clarity™ quality level = MQ100

430935-100G 430935-5G
SIAL430935-100GEA 12520 CZK
SIAL430935-100G SIAL430935-5G
PTFE, free flowing powder 1 µm, Sigma-Aldrich®
PTFE

Inert lubricant.


Low coefficient of friction, low critical surface energy and excellent antistick properties.

Vzorec: (CF₂CF₂)n
Hustota: 2.15 g/cm³ (25 °C)
MDL Number: MFCD00084455
CAS číslo: 9002-84-0

Objednat

Výsledky specifikačního testu

Appearance (colour) White to Off-White
Appearance (form) Powder
Infrared Spectrum Conforms to Structure
Size, Grind: 2.0 (NPIRI) (Maximum) Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR