Tisk…
α-D-Galacturonic acid monohydrate 97%
Dodavatel: Thermo Fisher Scientific
J66282.06 J66282.14
J66282.06EA 1750 CZK
J66282.06 J66282.14
α-D-Galacturonic acid monohydrate 97%
alpha-D-Galacturonic acid monohydrate
D-(+)-Galacturonic acid monohydrate can be used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
Vzorec: C₆H₁₀O₇·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00071585
CAS číslo: 91510-62-2
EINECS: 211-682-6
Merck Index: 14,04337