Tisk…
Capillary columns, OPTIMA® BioDiesel
Dodavatel: MACHEREY-NAGEL
726900.30 726903.10 726905.30
MANA726900.30EA 21890 CZK
MANA726900.30 MANA726903.10 MANA726905.30
Capillary columns, OPTIMA® BioDiesel
Kolony chromatografické
Special GC phases for the analysis of biodiesel.
  • OPTIMA® BioDiesel F for analysis of FAMEs in accordance with DIN EN 14103
  • OPTIMA® BioDiesel G for analysis of glycerol and glycerides in accordance with DIN EN 14105
  • OPTIMA® BioDiesel M for analysis of methanol in accordance with DIN EN 14110
The selectivities of the OPTIMA® BioDiesel capillary columns are specified for the analysis of quality parameters in biodiesel (DIN EN 14214 / ASTM D 6751).
Maximal temperature for isothermal operation/maximum temperature for short isotherms in a temperature program:
BioDiesel F: 240/250 °C
BioDiesel G: 380/400 °C
BioDiesel M: 340/360 °C