Tisk…

Citran železito-amonný (14,5 - 16,0% Fe) RT, green, powder, Purum p.a.

Dodavatel: Honeywell Chemicals

Synonyms: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

09713-1KG 09713-500G 09713-50G
09713-1KGEA 2440 CZK
09713-1KG 09713-500G 09713-50G
Citran železito-amonný (14,5 - 16,0% Fe) RT, green, powder, Purum p.a.
Citran železito-amonný

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Vzorec: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS číslo: 1185-57-5
EINECS: 214-686-6
Merck Index: 13,00518

Objednat

Výsledky specifikačního testu

Assay of Fe 14.5 - 16.0 %
Calcium (Ca) Max. 1000 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Potassium (K) Max. 500 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 100 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 500 ppm
Sulfate (SO₄) Max. 500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR