Tisk…
EDTA disodium salt concentrate for dilution to 1L, 0.1 M EDTA-Na2 Volumetric, Complexometry, Fluka™
Dodavatel: Honeywell Chemicals
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
38057-1EA 38057-6X1EA
38057-1EAEA 1100 CZK
38057-1EA 38057-6X1EA
EDTA disodium salt concentrate for dilution to 1L, 0.1 M EDTA-Na2 Volumetric, Complexometry, Fluka™
EDTA disodium salt
Concentrate for 1L standard solution
Vzorec: C₁₀H₁₄N₂Na₂O₈
MW: 336.21
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS číslo: 139-33-3
Merck Index: 12,03556
Objednat
Výsledky specifikačního testu
Factor 0.998 - 1.002