Tisk…

o-Xylén ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: 1,2-Dimethylbenzene

95662-250ML 95662-2.5L 95662-1L
95662-250MLEA 1420 CZK
95662-250ML 95662-2.5L 95662-1L
o-Xylén ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
o-Xylén

Beilstein Number: 1815558

Vzorec: C₈H₁₀
Bod varu: 7,6 %(V)
Bod tání: -25 °C
Hustota: 0,88 g/cm³ (20 °C)
Bod vzplanutí: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS číslo: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.504 - 1.506
Non-volatile matter Max. 0.002 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
m-, p-Xylene (GC) Max. 0.6 %
Toluene (GC) Max. 0.4 %
Explosion Limit 7 %
Vapor Density 3.7 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR