Poly(acrylic acid), M.W. 1080000 (MN 135000) secondary standard
Dodavatel: Thermo Fisher Scientific
178060100
178060010
ACRO178060100EA
8900
CZK
ACRO178060100
ACRO178060010
Poly(acrylic acid), M.W. 1080000 (MN 135000) secondary standard
Poly(acrylic acid)
Vzorec:
C₃H₄O₂ MW: 72.063 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00084394 CAS číslo: 9003-01-4 |
Výsledky specifikačního testu
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % |
Solubility | (2.5 % in pyridine) clear colorless to light yellow |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...