Taurodeoxycholic acid sodium salt hydrate ≥97% (by TLC), BioXtra, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N-(Deoxycholyl)taurine sodium salt, 2-([3|A,12|A-Dihydroxy-24-oxo-5|A-cholan-24-yl]amino)ethanesulphonic acid, Sodium 2-((R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulphonate xhydrate, Sodium taurodeoxycholate hydrate
M-Clarity™ quality level = MQ200
Sodium tauroursodeoxycholate is a bile salt which is naturally present in the small bowel.
- Aggregation number: 6
- CMC: one to four mM (20 to 25 °C)
- Solubility H2O: 0.5 M, clear, colorless to faintly yellow
Sodium tauroursodeoxycholates are anionic detergents used for isolation of membrane proteins including inner mitochondrial membrane proteins. These can be used in a study to assess short-term feedback regulation of bile salt uptake in rodent liver.
Can be used in a study to investigate its potential to resolve endoplasmic reticulum (ER) stress in intestinal epithelial cells.
Vzorec:
C₂₆H₄₄NNaO₆S.xH₂O MW: 521,69 g/mol (anhy) Bod tání: 168 °C (rozkl., lit.) |
MDL Number:
MFCD00149238 CAS číslo: 207737-97-1 |
Výsledky specifikačního testu
Assay | ≥97% (TLC) |
Phosphorus (P) | ≤0.002% |
Insoluble matter | ≤0.1% |
chloride (Cl-) | ≤0.5% |
sulfate (SO42-) | ≤0.05% |
Al | ≤0.0005% |
Ca | ≤0.005% |
Cu | ≤0.0005% |
Fe | ≤0.0005% |
K | ≤0.01% |
MG | ≤0.005% |
NH4+ | ≤0.05% |
PB | ≤0.001% |
Zn | ≤0.0005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...