Tisk…

Dioctyl ether 99% (by GC), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: n-Octyl ether, di-n-Octyl ether, Dioctylether

M-Clarity™ quality level = MQ100

249599-100G 249599-25G 249599-400G
SIAL249599-100GEA 4080 CZK
SIAL249599-100G SIAL249599-25G SIAL249599-400G
Dioctyl ether 99% (by GC), Sigma-Aldrich®
Dioctyl ether

Used in the preparation of11 nm sized monodisperse iron nanoparticles, cobalt ferrite nanocrystals with a particle diameter of 6 nm and CoPt-polymer nanostructured composites.

Vzorec: C₁₆H₃₄O
MW: 242.45 g/mol
Bod varu: 286...287 °C (lit.)
Bod tání: –7.6 °C (lit.)
Hustota: 0.806 g/ml (25 °C) (lit.)
MDL Number: MFCD00009563
CAS číslo: 629-82-3

Objednat

Výsledky specifikačního testu

Purity (GC) ≥98.5%
Infrared Spectrum Conforms to Structure
Refractive Index n20/D 1.433 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR