Tisk…
Transfection control plasmids, pVectOZ
Dodavatel: OZ BIOSCIENCES
PL00020 PL00120 PL00010 PL00110 PL00030 PL00130 PL00040 PL00140 PL00050 PL00150
OZBIPL00020EA 10030 CZK
OZBIPL00020 OZBIPL00120 OZBIPL00010 OZBIPL00110 OZBIPL00030 OZBIPL00130 OZBIPL00040 OZBIPL00140 OZBIPL00050 OZBIPL00150
Transfection control plasmids, pVectOZ
Vektory, plazmidy a knihovny
pVectOZ transfection plasmids are engineered in an optimised backbone and are ideal for all transfections.
  • Highest levels of transgene expression in mammalian cells and tissues
  • Suitable for all transfection applications (in vivo and in vitro)
  • High copy number production in Escherichia coli
  • LPS-endotoxin free, supercoiled and highly purified: transfection grade approved
  • Recommended for positive control and optimisation of all transfection experiments
Comprising five expression vectors encoding for the most popular reporter genes (CAT, GFP, LacZ, Luciferase, SEAP).
Informace o dodávce: Shipped at room temperature
Balení: Store at –20 °C.