1-Benzylpyrrole 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H55936.03
H55936.06
H55936.14
H55936.03EA
1100
CZK
H55936.03
H55936.06
H55936.14
1-Benzylpyrrole 96%
1-Benzylpyrrole
Vzorec:
C₁₁H₁₁N MW: 157.22 g/mol Bod varu: 90 °C (0.9 mmHg) Hustota: 1.020 Storage Temperature: Ambient |
MDL Number:
MFCD00963301 CAS číslo: 2051-97-0 EINECS: 218-137-1 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...