Tisk…

Methoxy(cyclooctadiene)rhodium(I) dimer, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

661058-1G 661058-250MG
SIAL661058-1GEA 43830 CZK
SIAL661058-1G SIAL661058-250MG
Methoxy(cyclooctadiene)rhodium(I) dimer, Sigma-Aldrich®
Methoxy(cyclooctadiene)rhodium(I) dimer

Application: Precursor for organorhodium(I) species, catalyst for cascade1,4-addition-aldol reaction, 1,4-addition of alcohols, and hydroformylation

Vzorec: C₁₈H₃₀O₂Rh₂
Bod tání: 192…225 °C
MDL Number: MFCD08457639
CAS číslo: 12148-72-0

Objednat

Výsledky specifikačního testu

Appearance (Color) Faint Yellow to Very Dark Yellow
Appearance (Form) Powder or Solid
Proton NMR spectra Conforms to Structure
Carbon 42.5-46.9 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR