Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan ≥97.0% (by HPLC), Sigma-Aldrich®
Dodavatel: Merck
47561-25G-F
47561-5G-F
47561-100G-F
47561-1KG-F
SIAL47561-25G-FEA
0
CZK
SIAL47561-25G-F
SIAL47561-5G-F
SIAL47561-100G-F
SIAL47561-1KG-F
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan ≥97.0% (by HPLC), Sigma-Aldrich®
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan
Vzorec:
C₃₁H₃₀N₂O₆ MW: 526,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153366 CAS číslo: 143824-78-6 |
Výsledky specifikačního testu
Purity (HPLC Area) | ≥97.0% |
Specific Rotation (20/D) | -21.0 ±2.0° |
Concentration | C=1 in DMF |
Proton NMR Spectrum | Conforms to Structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...