Tisk…

N-ɑ-Fmoc-N-Trityl-L-histidine ≥98.0% (by HPLC, sum of enantiomers), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-ɑ-Fmoc-tele-trityl-L-histidine, Fmoc-His(Trt)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-ɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-L-histidine, N-Fmoc-1-trityl-L-histidine

47639-5G-F 47639-25G-F 47639-100G-F
SIAL47639-5G-FEA 1710 CZK
SIAL47639-5G-F SIAL47639-25G-F SIAL47639-100G-F
N-ɑ-Fmoc-N-Trityl-L-histidine ≥98.0% (by HPLC, sum of enantiomers), Sigma-Aldrich®
N-alpha-Fmoc-N-Trityl-L-histidine
Vzorec: C₄₀H₃₃N₃O₄
MW: 619.72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00043332
CAS číslo: 109425-51-6

Objednat

Výsledky specifikačního testu

Assay (sum of enantiomers, HPLC) ≥98.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR