Tisk…

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine ≥98.0% (by HPLC), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Fmoc-Thr(tBu)-OH, Nalpha-Fmoc-O-tert-butyl-L-threonine, O-tert-Butyl-Nalpha-Fmoc-L-threonine, FMOC-O-tert-Butyl-L-threonine

47622-10G-F 47622-1G-F 47622-100G-F 47622-1KG-F 47622-25G-F
SIAL47622-10G-FEA 2470 CZK
SIAL47622-10G-F SIAL47622-1G-F SIAL47622-100G-F SIAL47622-1KG-F SIAL47622-25G-F
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine ≥98.0% (by HPLC), Sigma-Aldrich®
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine
Vzorec: C₂₃H₂₇NO₅
MW: 397,5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077075
CAS číslo: 71989-35-0

Objednat

Výsledky specifikačního testu

Assay (HPLC) ≥98%
Optical activity [α]20/D +16±1°, c = 1% in ethyl acetate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR