Tisk…

Nortriptyline hydrochloride ≥98%, crystalline solid

Dodavatel: Cayman Chemical
Varování

15904-10 15904-100 15904-50
CAYM15904-10EA 1320 CZK
CAYM15904-10 CAYM15904-100 CAYM15904-50
Nortriptyline hydrochloride ≥98%, crystalline solid
Nortriptyline hydrochloride

Nortriptyline is a tricyclic antidepressant that blocks norepinephrine and serotonin transporters (KDs = 4.4 and 18 nM, respectively) more potently than the dopamine transporter (KD = 1.1 µM). It also antagonises serotonin, histamine, muscarinic, and α-adrenergic receptors (Kis = 5.0, 8.5, 10, 40, and 60 nM for 5-HT2A, 5-HT2C, H1, α1, and M1 receptors, respectively).


Upozornení: Not for human or veterinary use.

Vzorec: C₁₉H₂₁N
Storage Temperature: Freezer
MDL Number: MFCD00058024
CAS číslo: 894-71-3

Objednat

Výsledky specifikačního testu

Purity ≥98%
λmax 240 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR