Tisk…

Guar gum, powder, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

G4129-250G G4129-1KG G4129-500G
SIALG4129-250GEA 1570 CZK
SIALG4129-250G SIALG4129-1KG SIALG4129-500G
Guar gum, powder, Sigma-Aldrich®
Guar gum

Guar gum is isolated from the endosperm of the guar or gavar bean.


Guar gum has been shown to have numerous beneficial effects including improved glucose tolerance and attenuation of diabetic nephropathy.

MDL Number: MFCD00131250
CAS číslo: 9000-30-0
EINECS: 232-536-8

Objednat

Výsledky specifikačního testu

Loss on drying ≤13%
Total ash ≤1%
pH 1% in H₂O (25 °C) 5.0 - 6.4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR