Tisk…

N-Fmoc-L-alanine 95% (by HPLC), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH

M-Clarity™ quality level = MQ100

531480-25G 531480-5G 531480-100G
SIAL531480-25GEA 1020 CZK
SIAL531480-25G SIAL531480-5G SIAL531480-100G
N-Fmoc-L-alanine 95% (by HPLC), Sigma-Aldrich®
N-Fmoc-L-alanine
Vzorec: C₁₈H₁₇NO₄
MW: 311.34 g/mol
Bod tání: 147…153 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00037139
CAS číslo: 35661-39-3
EINECS: 252-660-6

Objednat

Výsledky specifikačního testu

Infrared spectrum Conforms to structure
Proton NMR spectrum Conforms to structure
Optical rotation (C=1%, DMF) -20 to -16°
Purity (HPLC) ≥94.0%
Water (by Karl fischer) ≤6.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR