2-O-ɑ-D-glucopyranosyl-L-ascorbic acid 99.5%
Dodavatel: Thermo Scientific
Synonyms:
(R)-5-((S)-1,2-Dihydroxyethyl)-4-hydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)furan-2(5H)-one, AA-2G, Ascorbic acid 2-glucoside
J66601.03
J66601.06
J66601.03EA
1190
CZK
J66601.03
J66601.06
2-O-ɑ-D-glucopyranosyl-L-ascorbic acid 99.5%
2-O-alpha-D-glucopyranosyl-L-ascorbic acid
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduces the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. 2-O-alpha-D-Glucopyranosyl-L-ascorbic acid also promotes collagen synthesis and function as an antioxidant.
Vzorec:
C₁₂H₁₈O₁₁ MW: 338,27 g/mol Bod tání: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD23701380 CAS číslo: 129499-78-1 |
Výsledky specifikačního testu
Purity | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...