Lysostaphin (from Staphylococcus staphylolyticus)
Dodavatel: Merck
Lysostaphin is a zinc endopeptidase with a molecular weight of approximately 25 kDa. It contains 50 to 70% protein biuret.
- Units: ≥500 units/mg protein
- pH optimum for activity: ~7,5
Lysostaphin cleaves the polyglycine cross-links in the peptidoglycan layer of the cell wall of Staphylococcus species it has been found useful for cell lysis and also as a potential anti-microbial therapeutic.
Balení: Packed in glass bottle.