Tisk…
Lysostaphin (from Staphylococcus staphylolyticus)
Dodavatel: Merck
L7386-15MG L7386-5MG L7386-1MG
SIALL7386-15MGEA 27130 CZK
SIALL7386-15MG SIALL7386-5MG SIALL7386-1MG
Lysostaphin (from Staphylococcus staphylolyticus)
Proteiny
Lysostaphin is a zinc endopeptidase with a molecular weight of approximately 25 kDa. It contains 50 to 70% protein biuret.
  • Units: ≥500 units/mg protein
  • pH optimum for activity: ~7,5
Lysostaphin cleaves the polyglycine cross-links in the peptidoglycan layer of the cell wall of Staphylococcus species it has been found useful for cell lysis and also as a potential anti-microbial therapeutic.
Balení: Packed in glass bottle.