Tisk…

Taurolidine ≥98%, crystalline solid

Dodavatel: Cayman Chemical

15374-10 15374-5 15374-25 15374-50
CAYM15374-10EA 2240 CZK
CAYM15374-10 CAYM15374-5 CAYM15374-25 CAYM15374-50
Taurolidine ≥98%, crystalline solid
Taurolidine

Taurolidine is a synthetic taurine analog with antimicrobial and anti-neoplastic actions. These displays broad bactericidal and fungicidal activity.


Taurolidine and its metabolites mimic mannose by binding to the extracellular wall of bacteria, blocking adherence to epithelial and fibroblast cells. In addition to modulating immunoregulatory systems, taurolidine induces autophagy, apoptosis, and necrosis in human cancer cells. These produces cell death in malignant mesothelioma cells by oxidative stress in concert with inhibition of Akt signaling.

Vzorec: C₇H₁₆N₄O₄S₂
Storage Temperature: Freezer
MDL Number: MFCD00865076
CAS číslo: 19388-87-5

Objednat

Výsledky specifikačního testu

Purity ≥98%

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR