Tisk…

Acid red 52 ~75% (dye content), powder, BioReagent fluorescent dye, cell culture tested, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

S1402-25G S1402-5G S1402-1G
SIALS1402-25GEA 32220 CZK
SIALS1402-25G SIALS1402-5G SIALS1402-1G
Acid red 52 ~75% (dye content), powder, BioReagent fluorescent dye, cell culture tested, Sigma-Aldrich®
Acid red 52

Sulforhodamine B sodium salt used as a fluorescent dye to quantify cellular proteins in cultured cells.


  • fluorescence - λex 565 nm; λem 586 nm in H2O

Vzorec: C₂₇H₃₀N₂O₇S₂
MDL Number: MFCD00010180
CAS číslo: 3520-42-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR