Tisk…

N,N-Dimethylaminomethylferrocene 96% (by GC), dark brown liquid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ferrocenylmethyldimethylamine, (Dimethylaminomethyl)ferrocene

M-Clarity™ quality level = MQ100

119342-10G 119342-50G
SIAL119342-10GEA 2470 CZK
SIAL119342-10G SIAL119342-50G
N,N-Dimethylaminomethylferrocene 96% (by GC), dark brown liquid, Sigma-Aldrich®
N,N-Dimethylaminomethylferrocene
Vzorec: C₁₃H₁₇FeN
Bod varu: 124,..128 °C (2.5 mmHg)) (lit.)
Hustota: 1,228 g/ml (25 °C) (lit.)
Bod vzplanutí: 95 °C
MDL Number: MFCD00001433
CAS číslo: 1271-86-9
EINECS: 215-044-8

Objednat

Výsledky specifikačního testu

Proton NMR Spectrum Conforms to Structure
Purity (GC) ≥95.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR