Tisk…

Adamantane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

100277-100G 100277-25G
SIAL100277-100GEA 2450 CZK
SIAL100277-100G SIAL100277-25G
Adamantane, Sigma-Aldrich®
Adamantane
Vzorec: C₁₀H₁₆
MW: 136,24 g/mol
Bod varu: 171 °C (1013 hPa)
Bod tání: –25 °C
Hustota: 1,07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074719
CAS číslo: 281-23-2
EINECS: 206-001-4
Merck Index: 13,00149

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR