1-Chloroadamantane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
294861-25G
294861-5G
SIAL294861-25GEA
3290
CZK
SIAL294861-25G
SIAL294861-5G
1-Chloroadamantane, Sigma-Aldrich®
1-Chloroadamantane
Vzorec:
C₁₀H₁₅Cl MW: 170,68 g/mol Bod tání: 163…166 °C Hustota: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00075627 CAS číslo: 935-56-8 EINECS: 213-305-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...