Tisk…

1-Bromoadamantane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

109223-100G 109223-25G
SIAL109223-100GEA 4620 CZK
SIAL109223-100G SIAL109223-25G
1-Bromoadamantane, Sigma-Aldrich®
1-Bromoadamantane
Vzorec: C₁₀H₁₅Br
Bod varu: 226 °C (1013 hPa)
Bod tání: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00074721
CAS číslo: 768-90-1
EINECS: 212-200-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR