Symclosene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Trichloro-1,3,5-triazinetrion, 1,3,5-Trichlorohexahydro-1,3,5-triazine-2,4,6-trione, Trichlorocyanuric acid, TCCA, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, Trichloroisocyanuric acid
M-Clarity™ quality level = MQ100
91220-100G
91220-500G
SIAL91220-100GEA
723
CZK
SIAL91220-100G
SIAL91220-500G
Symclosene, Sigma-Aldrich®
Symclosene
Vzorec:
C₃Cl₃N₃O₃ MW: 232,41 g/mol Bod varu: 413 °C (1013 hPa) Bod tání: 225…230 °C Hustota: 2,07 g/cm³ (20 °C) Bod vzplanutí: >225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006553 CAS číslo: 87-90-1 EINECS: 201-782-8 UN: 2468 ADR: 5.1,II Merck Index: 12,09188 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...