Tisk…

Symclosene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Trichloro-1,3,5-triazinetrion, 1,3,5-Trichlorohexahydro-1,3,5-triazine-2,4,6-trione, Trichlorocyanuric acid, TCCA, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, Trichloroisocyanuric acid

M-Clarity™ quality level = MQ100

176125-1KG 176125-250G 176125-50G
SIAL176125-1KGEA 0 CZK
SIAL176125-1KG SIAL176125-250G SIAL176125-50G
Symclosene, Sigma-Aldrich®
Symclosene
Vzorec: C₃Cl₃N₃O₃
MW: 232,41 g/mol
Bod varu: 413 °C (1013 hPa)
Bod tání: 225…230 °C
Hustota: 2,07 g/cm³ (20 °C)
Bod vzplanutí: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00006553
CAS číslo: 87-90-1
EINECS: 201-782-8
UN: 2468
ADR: 5.1,II
Merck Index: 12,09188

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR