2-Acetylthiophene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Thiophen-2-yl-ethanone, 1-(2-Thienyl)-1-ethanone, Methyl 2-thienyl ketone
A13062.0E
A13062.22
A13062.36
A13062.0EEA
24440
CZK
A13062.0E
A13062.22
A13062.36
2-Acetylthiophene ≥99%
2-Acetylthiophene
Purity: 99%
CAS No.: 88-15-3
Molecular Formula: C6H6OS
Formula Weight: 126.18
Flash Point: 91°(195°F)
Refractive Index: 1.5660
Odour: Slightly sweetish
Melting Point: 10-12°
Boiling Point: 213-214°
Beilstein Reference: 107910
UN No.: 2810
MDL No.: MFCD00005442
Vzorec:
C₆H₆OS MW: 126,18 g/mol Bod varu: 213…214 °C Bod tání: 10…12 °C Hustota: 1,168 Bod vzplanutí: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005442 CAS číslo: 88-15-3 EINECS: 201-804-6 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...