2-Bromo-5-methylthiophene, Sigma-Aldrich®
Synonyms:
5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol
M-Clarity™ quality level = MQ200
515485-1G
515485-5G
SIAL515485-1G
SIAL515485-5G
2-Bromo-5-methylthiophene, Sigma-Aldrich®
2-Bromo-5-methylthiophene
Vzorec:
C₅H₅BrS MW: 177,06 g/mol Bod varu: 178 °C (1013 hPa) Bod tání: 58 °C Hustota: 1,5529 g/cm³ (20 °C) |
MDL Number:
MFCD00130103 CAS číslo: 765-58-2 EINECS: 212-151-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...