2-Thiophenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Thiophene-2-sulphonyl chloride
M-Clarity™ quality level = MQ200
255335-25G
255335-5G
SIAL255335-25GEA
0
CZK
SIAL255335-25G
SIAL255335-5G
2-Thiophenesulphonyl chloride, Sigma-Aldrich®
2-Thiophenesulphonyl chloride
Vzorec:
C₄H₃ClO₂S₂ MW: 182,65 g/mol Bod varu: 82…84 °C (1 mmHg) Bod tání: 32 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005426 CAS číslo: 16629-19-9 EINECS: 240-677-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...