2-Thiophenesulphonyl chloride ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15553.06
A15553.14
A15553.06EA
2160
CZK
A15553.06
A15553.14
2-Thiophenesulphonyl chloride ≥97%
2-Thiophenesulphonyl chloride
Purity: 97%
CAS No.: 16629-19-9
Molecular Formula: C4H3ClO2S2
Formula Weight: 182.65
Flash Point:
Vzorec:
C₄H₃ClO₂S₂ Bod varu: 80…81 °C (1 mmHg) Bod tání: 29…31 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005426 CAS číslo: 16629-19-9 EINECS: 240-677-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...