Tisk…

2,5-Diiodothiophene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

331570-1G 331570-5G
SIAL331570-1GEA 1570 CZK
SIAL331570-1G SIAL331570-5G
2,5-Diiodothiophene, Sigma-Aldrich®
2,5-Diiodothiophene
Vzorec: C₄H₂I₂S
MW: 335,93 g/mol
Bod varu: 67…70 °C (0,05 mmHg)
Bod tání: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014525
CAS číslo: 625-88-7
EINECS: 210-915-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR