2,5-Dichlorothiophene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
D75807-100G
D75807-25G
SIALD75807-100GEA
4560
CZK
SIALD75807-100G
SIALD75807-25G
2,5-Dichlorothiophene, Sigma-Aldrich®
2,5-Dichlorothiophene
Vzorec:
C₄H₂Cl₂S MW: 153,03 g/mol Bod varu: 162 °C (1013 hPa) Bod tání: –40 °C Hustota: 1,4473 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005423 CAS číslo: 3172-52-9 EINECS: 221-638-8 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...