Tisk…

2-Bromo-5-nitrothiophene ≥97%

Dodavatel: Thermo Scientific
Varování

A15866.06 A15866.14
A15866.06EA 4260 CZK
A15866.06 A15866.14
2-Bromo-5-nitrothiophene ≥97%
2-Bromo-5-nitrothiophene

Purity: 97% CAS No.: 13195-50-1 Molecular Formula: C4H2BrNO2S Formula Weight: 208.04 Melting Point: 42-46° Boiling Point: 235-237° MDL No.: MFCD00022493

Vzorec: C₄H₂BrNO₂S
MW: 208,04 g/mol
Bod varu: 235…237 °C
Bod tání: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00022493
CAS číslo: 13195-50-1
EINECS: 236-155-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR