Tisk…

1,1,3,3,5,5-Hexamethyltrisiloxane ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L16520.14 L16520.22
L16520.14EA 2470 CZK
L16520.14 L16520.22
1,1,3,3,5,5-Hexamethyltrisiloxane ≥97%
1,1,3,3,5,5-Hexamethyltrisiloxane

Purity: 97% CAS No.: 1189-93-1 Molecular Formula: C6H20O2Si3 Formula Weight: 208.48 Flash Point: 20°(68°F) Refractive Index: 1.3810 Melting Point: -67° Boiling Point: 128° Beilstein Reference: 1746341 UN No.: 1993 MDL No.: MFCD00039788

Vzorec: C₆H₂₀O₂Si₃
Bod varu: 128 °C
Bod tání: –67 °C
Hustota: 0,822
Bod vzplanutí: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039788
CAS číslo: 1189-93-1
EINECS: 214-716-8
UN: 1993
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR