Pentamethyldisiloxane 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1,1,1,3,3-Pentamethyldisiloxane
L16577.06
L16577.14
L16577.06EA
1760
CZK
L16577.06
L16577.14
Pentamethyldisiloxane 95%
Pentamethyldisiloxane
Purity: 95%
CAS No.: 1438-82-0
Molecular Formula: (CH3)3SiOSiH(CH3)2
Formula Weight: 148.35
Flash Point: -19°(-2°F)
Refractive Index: 1.3740
Boiling Point: 86-87°
Beilstein Reference: 1734568
UN No.: 1993
MDL No.: MFCD00053658
Vzorec:
C₅H₁₆OSi₂ MW: 148,35 g/mol Bod varu: 86…87 °C Hustota: 0,758 Bod vzplanutí: –19 °C (–2 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053658 CAS číslo: 1438-82-0 EINECS: 215-873-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...