1,1,3,3-Tetramethyldisiloxane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
235733-100G
235733-25G
SIAL235733-100GEA
3230
CZK
SIAL235733-100G
SIAL235733-25G
1,1,3,3-Tetramethyldisiloxane, Sigma-Aldrich®
1,1,3,3-Tetramethyldisiloxane
Vzorec:
C₄H₁₄OSi₂ MW: 134.33 Bod varu: 71 Hustota: 0.7555 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008256 CAS číslo: 3277-26-7 EINECS: 221-906-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...