1,1,3,3-Tetramethyldisiloxane ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B23697.14
B23697.22
B23697.36
B23697.14EA
1460
CZK
B23697.14
B23697.22
B23697.36
1,1,3,3-Tetramethyldisiloxane ≥97%
1,1,3,3-Tetramethyldisiloxane
Purity: 97%
CAS No.: 3277-26-7
Molecular Formula: C4H14OSi2
Formula Weight: 134.33
Flash Point: -26°(-15°F)
Refractive Index: 1.3700
Odour: Characteristic
Melting Point: -78°
Boiling Point: 70-71°
Beilstein Reference: 1733364
UN No.: 1993
MDL No.: MFCD00008256
Vzorec:
C₄H₁₄OSi₂ MW: 134.33 Bod varu: 70…71 °C Bod tání: –78 °C Hustota: 0.760 Bod vzplanutí: –26 °C (–15 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008256 CAS číslo: 3277-26-7 EINECS: 221-906-4 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...