2-Acetyl-1-methyl-1H-pyrrole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
M-Clarity™ quality level = MQ100
160865-10G
160865-1G
SIAL160865-10GEA
3940
CZK
SIAL160865-10G
SIAL160865-1G
2-Acetyl-1-methyl-1H-pyrrole, Sigma-Aldrich®
2-Acetyl-1-methyl-1H-pyrrole
Vzorec:
C₇H₉NO MW: 123,15 g/mol Bod varu: 200 °C (1013 hPa) Hustota: 1,0445 g/cm³ (15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003089 CAS číslo: 932-16-1 EINECS: 213-247-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...