2-Acetyl-1-methyl-1H-pyrrole ≥98%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
A15353.06
A15353.14
A15353.22
A15353.06EA
1020
CZK
A15353.06
A15353.14
A15353.22
2-Acetyl-1-methyl-1H-pyrrole ≥98%
2-Acetyl-1-methyl-1H-pyrrole
Purity: 98%
CAS No.: 932-16-1
Molecular Formula: C7H9NO
Formula Weight: 123.16
Flash Point: 68°(154°F)
Refractive Index: 1.5420
Odour: Pleasant
Boiling Point: 200-202°
Beilstein Reference: 111887
MDL No.: MFCD00003089
Vzorec:
C₇H₉NO MW: 123,15 g/mol Bod varu: 200…202 °C Hustota: 1,04 Bod vzplanutí: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003089 CAS číslo: 932-16-1 EINECS: 213-247-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...