2-Acetyl-1H-pyrrole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole
M-Clarity™ quality level = MQ300
W320218-100G-K
W320218-1KG-K
W320218-5KG-K
SIALW320218-100G-KEA
9590
CZK
SIALW320218-100G-K
SIALW320218-1KG-K
SIALW320218-5KG-K
2-Acetyl-1H-pyrrole, Sigma-Aldrich®
2-Acetyl-1H-pyrrole
Vzorec:
C₆H₇NO MW: 109,13 g/mol Bod varu: 220 °C (1013 hPa) Bod tání: 63…65 °C |
MDL Number:
MFCD00005220 CAS číslo: 1072-83-9 EINECS: 214-016-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...