2-Acetyl-1H-pyrrole ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A14593.06
A14593.14
A14593.06EA
1490
CZK
A14593.06
A14593.14
2-Acetyl-1H-pyrrole ≥98%
2-Acetyl-1H-pyrrole
Purity: 98%
CAS No.: 1072-83-9
Molecular Formula: C6H7NO
Formula Weight: 109.13
Odour: Characteristic
Melting Point: 88-92°
Boiling Point: 220°
Beilstein Reference: 1882
MDL No.: MFCD00005220
Vzorec:
C₆H₇NO Bod varu: 220 °C Bod tání: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005220 CAS číslo: 1072-83-9 EINECS: 214-016-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...