5-Methylpyrimidine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
686948-1G
686948-250MG
SIAL686948-1GEA
13290
CZK
SIAL686948-1G
SIAL686948-250MG
5-Methylpyrimidine, Sigma-Aldrich®
5-Methylpyrimidine
Vzorec:
C₅H₆N₂ MW: 94,12 g/mol Bod varu: 153 °C (1013 hPa) Bod tání: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047386 CAS číslo: 2036-41-1 EINECS: 218-006-9 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...