Pyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
131695-1G
131695-25G
131695-5G
SIAL131695-1GEA
1150
CZK
SIAL131695-1G
SIAL131695-25G
SIAL131695-5G
Pyrimidine, Sigma-Aldrich®
Pyrimidine
Vzorec:
C₄H₄N₂ MW: 80,09 g/mol Bod varu: 123 °C (1013 hPa) Bod tání: 21 °C Hustota: 1,529 g/cm³ (20 °C) |
MDL Number:
MFCD00006059 CAS číslo: 289-95-2 EINECS: 206-026-0 UN: 1993 ADR: 3,III Merck Index: 12,08170 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...